Merck
  • Modeling the precise interaction of glioblastoma with human brain region-specific organoids.
Modeling the precise interaction of glioblastoma with human brain region-specific organoids.
iScience (2024-02-23)
Qi Fan, Hanze Wang, Tianyi Gu, Huihui Liu, Peng Deng, Bo Li, Hui Yang, Ying Mao, Zhicheng Shao
ABSTRACT
Glioblastoma is a highly aggressive malignant tumor of the central nervous system, but the interaction between glioblastoma and different types of neurons remains unclear. Here, we established a co-culture model in vitro using 3D printed molds with microchannels, in which glioblastoma organoids (GB), dorsal forebrain organoids (DO, mainly composed of excitatory neurons), and ventral forebrain organoids (VO, mainly composed of inhibitory neurons) were assembled. Our results indicate that DO has a greater impact on altered gene expression profiles of GB, resulting in increased invasive potential. GB cells preferentially invaded DO along axons, whereas this phenomenon was not observed in VO. Furthermore, GB cells selectively inhibited neurite outgrowth in DOs and reduced the expression of the vesicular GABA transporter (VGAT), leading to neuronal hyperexcitability. By revealing how glioblastoma interacts with brain cells, our study provides a more comprehensive understanding of this disease.
Sigma-Aldrich
LDN193189 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nkx2.1 Antibody, clone 8G7-G3-1, ascites fluid, clone 8G7-G3-1, Chemicon®
Sigma-Aldrich
SAG dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
IWP-2, ≥98% (HPLC)
Sigma-Aldrich
Anti-Sox6 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®