Merck
  • Gla-rich protein is a novel vitamin K-dependent protein present in serum that accumulates at sites of pathological calcifications.
Gla-rich protein is a novel vitamin K-dependent protein present in serum that accumulates at sites of pathological calcifications.
The American journal of pathology (2009-11-07)
Carla S B Viegas, Sofia Cavaco, Pedro L Neves, Ana Ferreira, Alexandre João, Matthew K Williamson, Paul A Price, M Leonor Cancela, Dina C Simes
ABSTRACT
Mineralization of soft tissues is an abnormal process that occurs in any body tissue and can greatly increase morbidity and mortality. Vitamin K-dependent (VKD) proteins play a crucial role in these processes; matrix Gla protein is considered one of the most relevant physiological inhibitors of soft tissue calcification know to date. Several studies have suggested that other, still unknown, VKD proteins might also be involved in soft tissue calcification pathologies. We have recently identified in sturgeon a new VKD protein, Gla-rich protein (GRP), which contains the highest ratio between number of Gla residues and size of the mature protein so far identified. Although mainly expressed in cartilaginous tissues of sturgeon, in rat GRP is present in both cartilage and bone. We now show that GRP is a circulating protein that is also expressed and accumulated in soft tissues of rats and humans, including the skin and vascular system in which, when affected by pathological calcifications, GRP accumulates at high levels at sites of mineral deposition, indicating an association with calcification processes. The high number of Gla residues and consequent mineral binding affinity properties strongly suggest that GRP may directly influence mineral formation, thereby playing a role in processes involving connective tissue mineralization.
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Sigma-Aldrich
TWEEN® 20, Molecular Biology, viscous liquid
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
TWEEN® 20, viscosity 250-450 mPa.s (25 °C)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Chondroitinase ABC from Proteus vulgaris, BSA free, lyophilized powder, specific activity 50-250 units/mg protein
Sigma-Aldrich
Chondroitinase ABC from Proteus vulgaris, lyophilized powder, 0.3-3 units/mg solid
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid