Merck
  • Preclinical development of a stabilized RH5 virus-like particle vaccine that induces improved antimalarial antibodies.
Preclinical development of a stabilized RH5 virus-like particle vaccine that induces improved antimalarial antibodies.
Cell reports. Medicine (2024-07-18)
Lloyd D W King, David Pulido, Jordan R Barrett, Hannah Davies, Doris Quinkert, Amelia M Lias, Sarah E Silk, David J Pattinson, Ababacar Diouf, Barnabas G Williams, Kirsty McHugh, Ana Rodrigues, Cassandra A Rigby, Veronica Strazza, Jonathan Suurbaar, Chloe Rees-Spear, Rebecca A Dabbs, Andrew S Ishizuka, Yu Zhou, Gaurav Gupta, Jing Jin, Yuanyuan Li, Cecilia Carnrot, Angela M Minassian, Ivan Campeotto, Sarel J Fleishman, Amy R Noe, Randall S MacGill, C Richter King, Ashley J Birkett, Lorraine A Soisson, Carole A Long, Kazutoyo Miura, Rebecca Ashfield, Katherine Skinner, Mark R Howarth, Sumi Biswas, Simon J Draper
ABSTRACT
Plasmodium falciparum reticulocyte-binding protein homolog 5 (RH5) is a leading blood-stage malaria vaccine antigen target, currently in a phase 2b clinical trial as a full-length soluble protein/adjuvant vaccine candidate called RH5.1/Matrix-M. We identify that disordered regions of the full-length RH5 molecule induce non-growth inhibitory antibodies in human vaccinees and that a re-engineered and stabilized immunogen (including just the alpha-helical core of RH5) induces a qualitatively superior growth inhibitory antibody response in rats vaccinated with this protein formulated in Matrix-M adjuvant. In parallel, bioconjugation of this immunogen, termed "RH5.2," to hepatitis B surface antigen virus-like particles (VLPs) using the "plug-and-display" SpyTag-SpyCatcher platform technology also enables superior quantitative antibody immunogenicity over soluble protein/adjuvant in vaccinated mice and rats. These studies identify a blood-stage malaria vaccine candidate that may improve upon the current leading soluble protein vaccine candidate RH5.1/Matrix-M. The RH5.2-VLP/Matrix-M vaccine candidate is now under evaluation in phase 1a/b clinical trials.
Sigma-Aldrich
Anti-Rat IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution