Merck
  • Combined 3D and hypoxic culture improves cartilage-specific gene expression in human chondrocytes.
Combined 3D and hypoxic culture improves cartilage-specific gene expression in human chondrocytes.
Acta orthopaedica (2011-03-26)
Casper B Foldager, Anna B Nielsen, Samir Munir, Michael Ulrich-Vinther, Kjeld Søballe, Cody Bünger, Martin Lind
ABSTRACT
In vitro expansion of autologous chondrocytes is an essential part of many clinically used cartilage repair treatments. Native chondrocytes reside in a 3-dimensional (3D) network and are exposed to low levels of oxygen. We compared monolayer culture to combined 3D and hypoxic culture using quantitative gene expression analysis. Cartilage biopsies were collected from the intercondylar groove in the distal femur from 12 patients with healthy cartilage. Cells were used for either monolayer or scaffold culture. The scaffolds were clinically available MPEG-PLGA scaffolds (ASEED). After harvesting of cells for baseline investigation, the remainder was divided into 3 groups for incubation in conditions of normoxia (21% oxygen), hypoxia (5% oxygen), or severe hypoxia (1% oxygen). RNA extractions were performed 1, 2, and 6 days after the baseline time point, respectively. Quantitative RT-PCR was performed using assays for RNA encoding collagen types 1 and 2, aggrecan, sox9, ankyrin repeat domain-37, and glyceraldehyde-3-phosphate dehydrogenase relative to 2 hypoxia-stable housekeeping genes. Sox9, aggrecan, and collagen type 2 RNA expression increased with reduced oxygen. On day 6, the expression of collagen type 2 and aggrecan RNA was higher in 3D culture than in monolayer culture. Our findings suggest that there was a combined positive effect of 3D culture and hypoxia on cartilage-specific gene expression. The positive effects of 3D culture alone were not detected until day 6, suggesting that seeding of chondrocytes onto a scaffold for matrix-assisted chondrocyte implantation should be performed earlier than 2 days before implantation.
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Gentamicin solution, Hybri-Max, 50 mg/mL gentamicin, 0.1 μm filtered, BioReagent, suitable for hybridoma
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Gentamicin solution, 10 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Hyaluronidase from Streptomyces hyalurolyticus
Sigma-Aldrich
Penicillin-Streptomycin, Solution Stabilized, with 5,000 units penicillin and 5mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 10 purifications
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 350 purifications
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 70 purifications
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture