Merck
  • Establishing a 3D culture system for early organogenesis of monkey embryos ex vivo and single-cell transcriptome analysis of cultured embryos.
Establishing a 3D culture system for early organogenesis of monkey embryos ex vivo and single-cell transcriptome analysis of cultured embryos.
STAR protocols (2024-01-15)
Nianqin Sun, Honglian Shao, Youyue Zhang, Baiquan Ci, Hui Yao, Bing Bai, Tao Tan
ABSTRACT
Creating in vitro culture platforms for monkey embryos is crucial for understanding the initial 4 weeks of early primate embryogenesis. Here, we present a protocol to culture cynomolgus monkey embryos in vitro for 25 days post-fertilization and to delineate the key developmental events of gastrulation and early organogenesis. We describe steps for culturing with a 3D system, immunofluorescence analysis, single-cell RNA sequencing, and bioinformatic analysis. For complete details on the use and execution of this protocol, please refer to Gong et al. (2023).1.
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, from non-animal source, suitable for cell culture, meets EP, USP testing specifications
Sigma-Aldrich
HEPES, ≥99.5% (titration)