Merck
  • VTX-2337 is a novel TLR8 agonist that activates NK cells and augments ADCC.
VTX-2337 is a novel TLR8 agonist that activates NK cells and augments ADCC.
Clinical cancer research : an official journal of the American Association for Cancer Research (2011-12-01)
Hailing Lu, Gregory N Dietsch, Maura-Ann H Matthews, Yi Yang, Smita Ghanekar, Margaret Inokuma, Maria Suni, Vernon C Maino, Katherine E Henderson, James Jeffry Howbert, Mary L Disis, Robert M Hershberg
ABSTRACT
We aim to characterize VTX-2337, a novel Toll-like receptor (TLR) 8 agonist in clinical development, and investigate its potential to improve monoclonal antibody-based immunotherapy that includes the activation of natural killer (NK) cells. HEK-TLR transfectants were used to compare the selectivity and potency of VTX-2337, imiquimod, CpG ODN2006, and CL075. The ability of VTX-2337 to induce cytokine and chemokine production from human peripheral blood mononuclear cells (PBMC) and activation of specific immune cell subsets was examined. The potential for VTX-2337 to activate NK cell activity through direct and indirect mechanisms was also investigated. Finally, we tested the potential for VTX-2337 to augment antibody-dependent cell-mediated cytotoxicity (ADCC), especially in individuals with low-affinity FcγR3A single-nucleotide polymorphism (SNP). VTX-2337 selectively activates TLR8 with an EC(50) of about 100 nmol/L and stimulates production of TNFα and interleukin (IL)-12 from monocytes and myeloid dendritic cells (mDC). VTX-2337 stimulates IFNγ production from NK cells and increases the cytotoxicity of NK cells against K562 and ADCC by rituximab and trastuzumab. Effects of VTX-2337 on NK cells were, in part, from direct activation as increased IFNγ production and cytotoxic activity were seen with purified NK cells. Finally, VTX-2337 augments ADCC by rituximab in PBMCs with different FcγR3A genotypes (V/V, V/F, and F/F at position 158). VTX-2337 is a novel small-molecule TLR8 agonist that activates monocytes, DCs, and NK cells. Through the activation of NK cells, it has the potential to augment the effectiveness of monoclonal antibody treatments where a polymorphism in FcγR3A limits clinical efficacy.
Product Description
Motolimod, ≥98% (HPLC)