Merck
  • Sulfonation of papain-treated chitosan and its mechanism for anticoagulant activity.
Sulfonation of papain-treated chitosan and its mechanism for anticoagulant activity.
Carbohydrate research (2009-05-30)
Jiraporn Suwan, Zhenqing Zhang, Boyangzi Li, Preeyanat Vongchan, Puttinan Meepowpan, Fuming Zhang, Shaker A Mousa, Shaymaa Mousa, Bhusana Premanode, Prachya Kongtawelert, Robert J Linhardt
ABSTRACT
The novel low-molecular-weight chitosan polysulfate (MW 5120-26,200 Da) was prepared using the depolymerization of chitosan with papain (EC. 3.4.22.2). The sulfonation of depolymerized products was performed using chlorosulfonic acid in N,N-dimethylformamide under semi-heterogeneous conditions. The structures of the products were characterized by FTIR, (13)C NMR, and (1)H NMR (1D, 2D NMR) spectroscopy. The present study sheds light on the mechanism of anticoagulant activity of chitosan polysulfate. Anticoagulant activity was investigated by an activated partial thromboplastin assay, a thrombin time assay, a prothrombin time assay, and thrombelastography. Surface plasmon resonance also provided valuable data for understanding the relationship between the molecular binding of sulfated chitosan to two important blood clotting regulators, antithrombin III and heparin cofactor II. These results show that the principal mechanism by which this chitosan polysulfate exhibits anticoagulant activity is mediated through heparin cofactor II and is dependent on polysaccharide molecular weight.