Merck
  • Pathogenic MOG-reactive CD8+ T cells require MOG-reactive CD4+ T cells for sustained CNS inflammation during chronic EAE.
Pathogenic MOG-reactive CD8+ T cells require MOG-reactive CD4+ T cells for sustained CNS inflammation during chronic EAE.
Journal of neuroimmunology (2009-06-23)
Maria Bettini, Kristen Rosenthal, Brian D Evavold
ABSTRACT
XIncreasing evidence supports a role for CD8+ T cells in multiple sclerosis. In an attempt to isolate the contribution of CD8+ T cells in a murine model of MS, we immunized mice with a dominant CD8 epitope MOG37-46, a truncated version of MOG35-55. The data presented here show mild disease induced with MOG37-46, characterized by lower clinical scores, a decrease in CNS infiltration and a decrease in microglial activation. CD8+ T cells reactive to MOG37-46 are pro-inflammatory and traffic to the CNS; however, the presence of CD4+ T cells elicits more severe disease and sustained inflammation of the CNS.
Sigma-Aldrich
Avidin–Alkaline Phosphatase, buffered aqueous solution
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
Percoll®, suitable for plant cell culture
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)