Merck
  • Phase I and Pharmacology Study of Ropidoxuridine (IPdR) as Prodrug for Iododeoxyuridine-Mediated Tumor Radiosensitization in Advanced GI Cancer Undergoing Radiation.
Phase I and Pharmacology Study of Ropidoxuridine (IPdR) as Prodrug for Iododeoxyuridine-Mediated Tumor Radiosensitization in Advanced GI Cancer Undergoing Radiation.
Clinical cancer research : an official journal of the American Association for Cancer Research (2019-07-25)
Timothy Kinsella, Howard Safran, Susan Wiersma, Thomas DiPetrillo, Andrew Schumacher, Kayla Rosati, John Vatkevich, Lawrence W Anderson, Kimberly D Hill, Charles Kunos, Jerry M Collins
ABSTRACT
Iododeoxyuridine (IUdR) is a potent radiosensitizer; however, its clinical utility is limited by dose-limiting systemic toxicities and the need for prolonged continuous infusion. 5-Iodo-2-pyrimidinone-2'-deoxyribose (IPdR) is an oral prodrug of IUdR that, compared with IUdR, is easier to administer and less toxic, with a more favorable therapeutic index in preclinical studies. Here, we report the clinical and pharmacologic results of a first-in-human phase I dose escalation study of IPdR + concurrent radiation therapy (RT) in patients with advanced metastatic gastrointestinal (GI) cancers. Adult patients with metastatic GI cancers referred for palliative RT to the chest, abdomen, or pelvis were eligible for study. Patients received IPdR orally once every day × 28 days beginning 7 days before the initiation of RT (37.5 Gy in 2.5 Gy × 15 fractions). A 2-part dose escalation scheme was used, pharmacokinetic studies were performed at multiple time points, and all patients were assessed for toxicity and response to Day 56. Nineteen patients were entered on study. Dose-limiting toxicity was encountered at 1,800 mg every day, and the recommended phase II dose is 1,200 mg every day. Pharmacokinetic analyses demonstrated achievable and sustainable levels of plasma IUdR ≥1 μmol/L (levels previously shown to mediate radiosensitization). Two complete, 3 partial, and 9 stable responses were achieved in target lesions. Administration of IPdR orally every day × 28 days with RT is feasible and tolerable at doses that produce plasma IUdR levels ≥1 μmol/L. These results support the investigation of IPdR + RT in phase II studies.
Product Description
Ropidoxuridine, ≥98% (HPLC), powder, radiosensitizer