Merck
  • SUMOylation mediates the nuclear translocation and signaling of the IGF-1 receptor.
SUMOylation mediates the nuclear translocation and signaling of the IGF-1 receptor.
Science signaling (2010-02-11)
Bita Sehat, Ali Tofigh, Yingbo Lin, Eric Trocmé, Ulrika Liljedahl, Jens Lagergren, Olle Larsson
ABSTRACT
The insulin-like growth factor 1 receptor (IGF-1R) plays crucial roles in developmental and cancer biology. Most of its biological effects have been ascribed to its tyrosine kinase activity, which propagates signaling through the phosphatidylinositol 3-kinase and mitogen-activated protein kinase pathways. Here, we report that IGF-1 promotes the modification of IGF-1R by small ubiquitin-like modifier protein-1 (SUMO-1) and its translocation to the nucleus. Nuclear IGF-1R associated with enhancer-like elements and increased transcription in reporter assays. The SUMOylation sites of IGF-1R were identified as three evolutionarily conserved lysine residues-Lys(1025), Lys(1100), and Lys(1120)-in the beta subunit of the receptor. Mutation of these SUMO-1 sites abolished the ability of IGF-1R to translocate to the nucleus and activate transcription but did not alter its kinase-dependent signaling. Thus, we demonstrate a SUMOylation-mediated mechanism of IGF-1R signaling that has potential implications for gene regulation.
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block