Merck
  • A type II restriction endonuclease with an eight nucleotide specificity from Streptomyces fimbriatus.
A type II restriction endonuclease with an eight nucleotide specificity from Streptomyces fimbriatus.
Nucleic acids research (1984-06-11)
B Q Qiang, I Schildkraut
ABSTRACT
A new site-specific endonuclease, Sfi I, has been isolated from Streptomyces fimbriatus . This is the first report of a type II restriction endonuclease whose recognition specificity requires eight nucleotides. Sfi I cleaves the sequence, GGCCNNNN / NGGCC , symmetrically to produce a three base, 3' extension.