Merck
  • Isolation of a FAD-GPDH gene encoding a mitochondrial FAD-dependent glycerol-3-phosphate dehydrogenase from Dunaliella salina.
Isolation of a FAD-GPDH gene encoding a mitochondrial FAD-dependent glycerol-3-phosphate dehydrogenase from Dunaliella salina.
Journal of basic microbiology (2007-05-24)
Wanggui Yang, Yi Cao, Xiaofei Sun, Fei Huang, Qinghua He, Dairong Qiao, Linhan Bai
ABSTRACT
The mitochondrial FAD-dependent glycerol-3-phosphate dehydrogenase (FAD-GPDH), recently reported in plants, has been detailed in yeast and animal systems. It oxidizes glycerol-3-phosphate (G-3-P) to dihydroxyacetone phosphate (DHAP) on the outer surface of mitochondrial inner membrane. A cDNA encoding the Dunaliella salina mitochondrial glycerol-3-phosphate dehydrogenase (DsFAD-GPDH) has been cloned and sequenced. The full length cDNA is 2791 bp, with an open reading frame (ORF) encoding 650 predicted amino acids, which show strong homology to reported FAD-GPDHs and have an apparent mitochondrial targeting sequence in the N-terminal. The sequence has been submitted to the GenBank database under Accession No. DQ916107. Results of Real-Time Quantitative PCR and enzymatic assays show that expression of DsFAD-GPDH is enhanced at first by salt treatment, and repressed by oxygen deficiency and cold stress.
Sigma-Aldrich
Glycerol 3-phosphate Oxidase from Pediococcus sp., lyophilized powder, 40-80 units/mg solid, pH 8.1
Sigma-Aldrich
Glycerol 3-phosphate Oxidase from Aerococcus viridans, lyophilized powder, ≥70 units/mg solid