Merck
Reveals new lung adenocarcinoma cancer genes based on gene expression.
European review for medical and pharmacological sciences (2012-10-11)
Z-F Lin, X-Y Shen, F-Z Lu, Z Ruan, H-L Huang, J Zhen
ABSTRACT
Lung adenocarcinoma (LAC) is the most common type of lung cancer, accounting for 30-35% of all cases. In this study we aim to predict potential genes and confirm pathways which are associated with LAC. By using the meta-analysis method, GSE10072 and GSE 2514 datasets were merged to find potential genes and pathways which are associated with LAC. Our analysis indicated identified differentially expressed genes enriched in multicellular organismal metabolic process, gland development, and urogenital system development. Further, we predicted genes including EGF-like domain might be the potential target genes for further study, such as NGX6, MUC17, and Nel. In addition, a number of genes that associated with axon guidance, focal adhesion, and complement and coagulation cascades pathway might be also involved in LAC in a direct or indirectly manner. Our analysis indicated identified differentially expressed genes enriched in multicellular organismal metabolic process, gland development, and urogenital system development We anticipate numerous advances in LAC research in the coming years based on our meta-analysis.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
β-Mannosidase from Helix pomatia, 5-30 units/mL, ammonium sulfate suspension, crude extract
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid