Merck
The role of hyaluronic acid in SEB-induced acute lung inflammation.
Clinical immunology (Orlando, Fla.) (2012-12-19)
Olga N Uchakina, Clara M Castillejo, Christy C Bridges, Robert J McKallip
ABSTRACT
We investigated the role of the extracellular matrix component, hyaluronic acid (HA) in SEB-induced ALI/ARDS. Intranasal exposure of mice to SEB led to a significant increase in the level of soluble hyaluronic acid in the lungs. Similarly, in an endothelial cell/spleen cell co-culture, SEB exposure led to significant increases in soluble levels of hyaluronic acid, cellular proliferation, and cytokine production compared with SEB-exposed spleen cells or endothelial cells alone. Exposure of SEB-activated spleen cells to hyaluronic acid led to increased cellular proliferation and increased cytokine production. SEB-induced cytokine production and proliferation in vitro were significantly reduced by the hyaluronic acid blocking peptide, Pep-1. Finally, treatment of SEB-exposed mice with Pep-1 significantly reduced SEB-induced ALI/ARDS, through reduction of cytokine production and numbers of lung inflammatory cells, compared to mice treated with a control peptide. Together, these results suggest the possibility of targeting HA for the treatment of SEB-induced ALI/ARDS.
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg