Merck
  • Autocrine motility factor promotes HER2 cleavage and signaling in breast cancer cells.
Autocrine motility factor promotes HER2 cleavage and signaling in breast cancer cells.
Cancer research (2012-12-19)
Dhong Hyo Kho, Pratima Nangia-Makker, Vitaly Balan, Victor Hogan, Larry Tait, Yi Wang, Avraham Raz
ABSTRACT
Trastuzumab (Herceptin) is an effective targeted therapy in HER2-overexpressing human breast carcinoma. However, many HER2-positive patients initially or eventually become resistant to this treatment, so elucidating mechanisms of trastuzumab resistance that emerge in breast carcinoma cells is clinically important. Here, we show that autocrine motility factor (AMF) binds to HER2 and induces cleavage to the ectodomain-deleted and constitutively active form p95HER2. Mechanistic investigations indicated that interaction of AMF with HER2 triggers HER2 phosphorylation and metalloprotease-mediated ectodomain shedding, activating phosphoinositide-3-kinase (PI3K) and mitogen-activated protein kinase signaling and ablating the ability of trastuzumab to inhibit breast carcinoma cell growth. Furthermore, we found that HER2 expression and AMF secretion were inversely related in breast carcinoma cells. On the basis of this evidence that AMF may contribute to HER2-mediated breast cancer progression, our findings suggest that AMF-HER2 interaction might be a novel target for therapeutic management of patients with breast cancer, whose disease is resistant to trastuzumab.
Product Description
Supelco
Phosphoglucose Isomerase from baker′s yeast (S. cerevisiae), for use with Fructose Assay Kit FA-20
Sigma-Aldrich
Phosphoglucose Isomerase from Bacillus stearothermophilus, lyophilized powder, 300-1,000 units/mg protein
Sigma-Aldrich
Phosphoglucose Isomerase from rabbit muscle, Type XI, lyophilized powder, ≥200 units/mg protein
Sigma-Aldrich
Phosphoglucose Isomerase from baker′s yeast (S. cerevisiae), Type III, ammonium sulfate suspension, ≥400 units/mg protein (biuret)