Merck
  • Characterization of tyrosinase inhibitors in the twigs of Cudrania tricuspidata and their structure-activity relationship study.
Characterization of tyrosinase inhibitors in the twigs of Cudrania tricuspidata and their structure-activity relationship study.
Fitoterapia (2012-12-25)
Zong-Ping Zheng, Hui-Yuan Tan, Jie Chen, Mingfu Wang
ABSTRACT
The twigs of Cudrania tricuspidata were found to show strong tyrosinase inhibitory activity, and further detailed component analysis resulted in the isolation of a new flavanol glucoside, (2S,3S)-2,3-trans-dihydromorin-7-O-β-d-glucoside (1), plus twenty-seven known compounds (2-28). Their structures were elucidated on the basis of ESI-MS and NMR spectral data. Among the isolated compounds, trans-dihydromorin (8), oxyresveratrol (9), and steppogenin (12) were found to exhibit significant tyrosinase inhibition activities. Moreover, the structure-activity relationship of these isolated compounds was also discussed.
Sigma-Aldrich
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid