Merck
  • Trypsin and chymotrypsin inhibitor peptides from the venom of Chinese Daboia russellii siamensis.
Trypsin and chymotrypsin inhibitor peptides from the venom of Chinese Daboia russellii siamensis.
Toxicon : official journal of the International Society on Toxinology (2013-01-05)
Chun-teng Guo, Stephen McClean, Chris Shaw, Ping-fan Rao, Ming-yu Ye, Anthony J Bjourson
ABSTRACT
Two trypsin inhibitors and one chymotrypsin inhibitor from Chinese Daboia russellii siamensis venom, denoted as CBPTI-1, CBPTI-2 and CBPTI-3 were purified, characterized and cloned from lyophilized venom-derived cDNA libraries. The N-terminus of CBPTI-1 was modified and not amenable to Edman degradation sequencing, however an internal partial sequence was found to be SGRCRGHLRRIYYNPDSNKCE. The N-termini of CBPTI-2 and CBPTI-3 were unmodified and their partial sequences were established as HDRPTFCNLAPESGRCRAH and HDRPKFCYLPADPGECMAYIRSFYYDS respectively. From cloning studies CBPTI-1 was found to consist of 66 amino acid residues, while CBPTI-2 and CBPTI-3 precursors consist of 60 amino acid residues, including 6 cysteine residues. Another cDNA sequence (CBPTI-4) was also obtained. Alignment of cDNA sequences showed that CBPTI-3 exhibited similar sequence homology to CBPTI-4 cDNA except for an 8 nucleotide deletion in the open-reading frame. CBPTI-1 and CBPTI-2 were demonstrated to be potent trypsin inhibitors, but were also shown to be effectively potent in chymotrypsin inhibition. The K(i) values of CBPTI-1 and CBPTI-2 for trypsin inhibition were 4.07 × 10(-7) M and 6.66 × 10(-7) M, respectively, and they were non-competitive in their activity. CBPTI-3 showed chymotrypsin inhibition activity with a K(i) value of 2.55 × 10(-9) M, but did not show trypsin inhibitor activity.
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from human pancreas, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin−Agarose from bovine pancreas, lyophilized powder, 2,000-3,500 units/g agarose (One ml gel will yield 65-120 units)