Merck
  • Fission yeast RecQ helicase Rqh1 is required for the maintenance of circular chromosomes.
Fission yeast RecQ helicase Rqh1 is required for the maintenance of circular chromosomes.
Molecular and cellular biology (2013-01-09)
Tomoko Nanbu, Katsunori Takahashi, Johanne M Murray, Naoya Hirata, Shinobu Ukimori, Mai Kanke, Hisao Masukata, Masashi Yukawa, Eiko Tsuchiya, Masaru Ueno
ABSTRACT
Protection of telomeres protein 1 (Pot1) binds to single-stranded telomere overhangs and protects chromosome ends. RecQ helicases regulate homologous recombination at multiple stages, including resection, strand displacement, and resolution. Fission yeast pot1 and RecQ helicase rqh1 double mutants are synthetically lethal, but the mechanism is not fully understood. Here, we show that the synthetic lethality of pot1Δ rqh1Δ double mutants is due to inappropriate homologous recombination, as it is suppressed by the deletion of rad51(+). The expression of Rad51 in the pot1Δ rqh1Δ rad51Δ triple mutant, which has circular chromosomes, is lethal. Reduction of the expression of Rqh1 in a pot1 disruptant with circular chromosomes caused chromosome missegregation, and this defect was partially suppressed by the deletion of rad51(+). Taken together, our results suggest that Rqh1 is required for the maintenance of circular chromosomes when homologous recombination is active. Crossovers between circular monomeric chromosomes generate dimers that cannot segregate properly in Escherichia coli. We propose that Rqh1 inhibits crossovers between circular monomeric chromosomes to suppress the generation of circular dimers.
Sigma-Aldrich
Topoisomerase I human, buffered aqueous glycerol solution, ≥2 units/μL