Merck
  • Bronchoalveolar lavage amylase is associated with risk factors for aspiration and predicts bacterial pneumonia.
Bronchoalveolar lavage amylase is associated with risk factors for aspiration and predicts bacterial pneumonia.
Critical care medicine (2013-01-15)
Curtis H Weiss, Farzad Moazed, David DiBardino, Mamta Swaroop, Richard G Wunderink
ABSTRACT
Aspiration of oropharyngeal or gastric contents into the respiratory tract leads to a spectrum of disorders with high morbidity. Aspiration is a diagnostic dilemma, because clinical characteristics and diagnostic tests are not effective predicting or confirming aspiration. We sought to determine whether α-amylase, a protein secreted by salivary glands and the pancreas, is elevated in bronchoalveolar lavage specimens in patients with clinical risk factors for aspiration and whether bronchoalveolar lavage amylase predicts bacterial pneumonia. Retrospective analysis. Five adult ICUs at a tertiary care urban medical center. Mechanically ventilated patients who underwent either bronchoscopic or nonbronchoscopic bronchoalveolar lavage within 72 hrs of endotracheal intubation between August 1, 2008 and June 30, 2010. A total of 296 bronchoalveolar lavage amylase results from 280 patients were included in the analysis, and 155 bronchoalveolar lavage amylase specimens were obtained from patients with at least one predefined preintubation risk factor (altered consciousness, swallowing dysfunction, difficult intubation, peri-intubation vomiting, or cardiac arrest). Bronchoalveolar lavage amylase concentration increased as the number of preintubation risk factors increased (p < 0.001). In addition, bronchoalveolar lavage amylase was elevated in patients with bacterial pneumonia (cfu/mL ≥ 10) (p < 0.001). The area under the receiver operator curve for the ability of bronchoalveolar lavage amylase to differentiate between positive and negative bronchoalveolar lavage culture was 0.67 (95% confidence interval, 0.60-0.75). The lower 95% confidence interval for bronchoalveolar lavage amylase in patients with at least one preintubation risk factor for aspiration was 125.9 units/L. In multivariate analysis, bronchoalveolar lavage amylase < 125 units/L was associated with significantly lower odds of bacterial pneumonia (odds ratio 0.39, 95% confidence interval 0.21-0.71, p = 0.002). Elevated bronchoalveolar lavage amylase is associated with risk factors for aspiration and may predict bacterial pneumonia. Bronchoalveolar lavage amylase may be useful as an early screening tool to guide management of patients suspected of aspiration.
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)