Merck
  • N-octyl-N-Arginine chitosan micelles as an oral delivery system of insulin.
N-octyl-N-Arginine chitosan micelles as an oral delivery system of insulin.
Journal of biomedical nanotechnology (2013-04-30)
Zhen-Hai Zhang, Sarra Abbad, Rui-Rui Pan, Ayman Y Waddad, Lu-Lu Hou, Hui-Xia Lv, Jian-Ping Zhou
ABSTRACT
N-octyl-N-Arginine chitosan (OACS) was synthesized in an attempt to combine the permeation enhancing effects of arginine-rich peptides and the drug loading capacity of the amphipathic polymers for insulin oral delivery. OACS self-assembled micelles of insulin were prepared by the conventional stirring technique, which were characterized by Dynamic light scattering, transmission electron microscopy and differential scanning calorimetry. Molecular docking by Discovery studio software confirmed that the interactions between OACS and insulin were mostly electrostatic in nature. In vitro, the result of the degradation experiment by enzyme showed that the OACS has a relative protective effect for insulin from proteolyses. Compared to the insulin solution, OACS micelles increased the Caco-2 cell's internalization by up to 22.3 folds. In vivo, the pharmacological activity PA% of series OACS-insulin micelles ranged from 7.7%-16.8%. Meanwhile by increasing arginine degree of the substitution both the uptake in Caco-2 cells and the hypoglycemic effect in diabetic rats were enhanced. Therefore, it is concluded that using arginine polymeric micelles for the enhancement of oral insulin delivery is a promising approach for the oral peptide delivery.
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Pepsin−Agarose from porcine gastric mucosa, lyophilized powder, ≥30 units/mg dry solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, tablet, 1 mg tablet
Sigma-Aldrich
TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Trypsin from porcine pancreas, ~1500 U/mg
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous
Sigma-Aldrich
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder