Merck
  • [Establishment of recombinant Lac Z reporter gene-transformed yeast cells for bioassay of androgen-like compounds in environment].
[Establishment of recombinant Lac Z reporter gene-transformed yeast cells for bioassay of androgen-like compounds in environment].
Xi bao yu fen zi mian yi xue za zhi = Chinese journal of cellular and molecular immunology (2013-05-07)
Weixing Dai, Chen Yang, Fangni Luo, Shanshan Li, Renhua Huang, Yangmei Xie, Feiyue Xue, Xiangming Li
ABSTRACT
To establish the yeast-based bioassay system for the androgenic endocrine disruptors in the environment. The recombinant gene-transformed yeast cells were constructed based on two episomal vectors. In the expression vector, the expression of androgen receptor (AR) gene was driven by 3-glyceraldehydephosphate dehydrogenase (GPD) promoter and the gene was fused to V5 epitope; in the reporter vector, the expression of the Lac Z gene as a report gene was under the control of the androgen response element (ARE). The vectors were transformed into yeast cells (W303-1A) to construct the yeast-based bioassay system. The sensitivity of this assay system was evaluated by the androgenic compounds (dihydrotestosterone, DHT and testosterone propionate, TP)and the specificity of this system by the estrogenic compounds (17β-estradiol, estriol, estrone, diethylstilbestrol and ethinyl estradiol). Both DHT and TP yielded a significant dose-effect model with the recombinant gene-transformed yeast cells, indicating that this yeast bioassay system has a high sensitivity. Moreover, the assay also exhibited a high specificity in that the yeast cells had no dose-effect relationship with the different concentrations of five estrogens (17β-estradiol, estriol, estrone, diethylstilbestrol and ethinyl estradiol). The recombinant yeast cells could be applied to screen for the androgen-like compounds in the environment.
Sigma-Aldrich
β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)
Sigma-Aldrich
β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein