Merck
  • The Rag4 glucose sensor is involved in the hypoxic induction of KlPDC1 gene expression in the yeast Kluyveromyces lactis.
The Rag4 glucose sensor is involved in the hypoxic induction of KlPDC1 gene expression in the yeast Kluyveromyces lactis.
Eukaryotic cell (2010-11-26)
C Micolonghi, M Wésolowski-Louvel, M M Bianchi
ABSTRACT
Kluyveromyces lactis is a yeast which cannot grow under strict anaerobiosis. To date, no factors responsible for oxygen sensing and oxygen-dependent regulation of metabolism have been identified. In this paper we present the identification of the glucose sensor Rag4 as a factor essential for oxygen-dependent regulation of the fermentative pathway.