Merck
  • Chemoenzymatic synthesis of hydrophobic glycoprotein: synthesis of saposin C carrying complex-type carbohydrate.
Chemoenzymatic synthesis of hydrophobic glycoprotein: synthesis of saposin C carrying complex-type carbohydrate.
The Journal of organic chemistry (2012-07-18)
Hironobu Hojo, Hiromasa Tanaka, Masashi Hagiwara, Yuya Asahina, Akiharu Ueki, Hidekazu Katayama, Yuko Nakahara, Azusa Yoneshige, Junko Matsuda, Yukishige Ito, Yoshiaki Nakahara
ABSTRACT
The complex-type N-linked octasaccharide oxazoline having LacNAc as the nonreducing end sugar was efficiently synthesized using the benzyl-protected LacNAc, mannose, and β-mannosyl GlcNAc units as key building blocks. To achieve a highly β-selective glycosylation with the LacNAc unit, the N-trichloroacetyl group was used for the protection of the amino group in the LacNAc unit. After complete assembly of these units and deprotection, the obtained free sugar was successfully derivatized into the corresponding sugar oxazoline. On the other hand, the N-acetylglucosaminylated saposin C, a hydrophobic lipid-binding protein, was chemically synthesized by the native chemical ligation reaction. On the basis of the previous results related to the synthesis of the nonglycosylated saposin C, the O-acyl isopeptide structure was introduced to the N-terminal peptide thioester carrying GlcNAc to improve its solubility toward aqueous organic solvents. The ligation reaction efficiently proceeded with the simultaneous O- to N-acyl shift at the O-acyl isopeptide moiety. After the removal of the cysteine-protecting group and folding, saposin C carrying GlcNAc was successfully obtained. The synthetic sugar oxazoline was then transferred to this glycoprotein using the mutant of endo-β-N-acetylglucosaminidase from Mucor hiemalis (Endo-M) (glycosynthase), and the saposin C carrying the complex-type nonasaccharide was successfully obtained.
Sigma-Aldrich
Endoglycosidase F3 from Elizabethkingia miricola, recombinant, expressed in E. coli, 30 U/mg
Sigma-Aldrich
Endoglycosidase H from Streptomyces plicatus, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Endoglycosidase F2 from Elizabethkingia miricola, recombinant, expressed in E. coli, 20 U/mg
Sigma-Aldrich
Endoglycosidase F1 from Elizabethkingia miricola, recombinant, expressed in E. coli, ≥16 U/mg, buffered aqueous solution