Merck
  • Digestive enzyme as benchmark for insecticide resistance development in Culex pipiens larvae to chemical and bacteriologic insecticides.
Digestive enzyme as benchmark for insecticide resistance development in Culex pipiens larvae to chemical and bacteriologic insecticides.
Journal of the Egyptian Society of Parasitology (2013-05-24)
Nashwa H Kamel, Iman M Bahgat, Gamal A El Kady
ABSTRACT
This work monitored changes in some digestive enzymes (trypsin and aminopeptidase) associated with the building up of resistance in Cx. pipiens larvae to two chemical insecticides (methomyl and/or malathion) and one biological insecticide (Bacillus thuringiensis-H14 or B.t H 14). The LC50 value of methomyl for both field- and the 12th generation (F12) of the selected strain was 1.789 ppm and 8.925 ppm respectively. The LC50 value of malathion for both field and the F12 of the selected strain was 0.082 ppm and 0.156 ppm respectively, and those of B.t H14 of field strain and the F12 was 2.550ppm & 2.395ppm respectively. The specific activity of trypsin enzyme in control susceptible colony was 20.806 +/- 0.452micromol/min/mg protein; but at F4 and F8 for malathion and methomyl treated larvae were 10.810 +/- 0.860 & 15.616+/-0.408 micromol/min/mg protein, respectively. Trypsin activity of F12 in treated larvae with B.t.H14 was 2.097 +/- 0.587 microiol/min/mg protein. Aminopeptidase specific activity for susceptible control larvae was 173.05 +/- 1.3111 micromol/min/mg protein. This activity decreased to 145.15 +/- 4.12, 152.497 +/- 6.775 & 102.04 +/- 3.58a micromol/min/mg protein after larval (F 12) treatment with methomyl, malathion and B.t H 14 respectively.
Product Description
Supelco
Malathion, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Malathion solution, 100 μg/mL in cyclohexane, PESTANAL®, analytical standard
Supelco
Malathion, PESTANAL®, analytical standard
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, tablet, 1 mg tablet
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Trypsin from porcine pancreas, ~1500 U/mg
Supelco
Methomyl, PESTANAL®, analytical standard