Merck
  • Regulation of 5-hydroxytryptamine2 (5-HT2) receptor expression in cultured rat aortic smooth muscle cells by SR 46349B, a selective 5-HT2 receptor antagonist.
Regulation of 5-hydroxytryptamine2 (5-HT2) receptor expression in cultured rat aortic smooth muscle cells by SR 46349B, a selective 5-HT2 receptor antagonist.
The Journal of biological chemistry (1994-01-07)
M Rinaldi-Carmona, V Prabonnaud, M Bouaboula, C Poinot-Chazel, P Casellas, G Le Fur, J M Herbert
ABSTRACT
Regulation of 5-hydroxytryptamine (5-HT2) receptor expression by SR 46349B, a potent and selective 5-HT2 receptor antagonist, was investigated in cultured rat aortic smooth muscle cells. Binding of [3H]SR 46349B to rat vascular smooth muscle cells was time-dependent, reversible, and saturable. [3H]SR 46349B bound to one class of specific binding sites with high affinity (KD = 1.3 +/- 0.3 nM; Bmax = 176 +/- 42 fmol/10(5) cells). Exposure of cells to a 1 microM concentration of the 5-HT2 agonist (+/-)-1-(2,5-dimethoxy-4-iodophenyl)-2-aminopropane ((+/-)-DOI) or the antagonist ketanserin led to a significant decrease in 5-HT2 receptor density as measured by [3H]SR 46349B binding. In contrast, exposure of cells to 1 microM SR 46349B caused a marked increase in the maximal binding capacity of [3H]SR 46349B, with a maximal effect at 24 h (73% increase). The affinity constant was not affected by prior exposure to (+/-)-DOI, ketanserin, or SR 46349B. Furthermore, exposure of cells to 1 microM (+/-)-DOI or ketanserin produced, 48 h later, a decrease in the ability of (+/-)-DOI to stimulate phosphoinositide turnover in the cells, whereas treatment with SR 46349B induced a significant stimulation of the 5-HT2 receptor-linked signal transduction. This effect occurred with no changes in the amount of 5-HT2 receptor mRNAs as measured by quantitative polymerase chain reaction. These results indicate that SR 46349B increases 5-HT2 receptor binding and functions without altering steady-state 5-HT2 mRNA levels in cultured rat aortic smooth muscle cells.
Sigma-Aldrich
SR-46349 hemifumarate salt, ≥98% (HPLC)