Merck
  • Mutations of the human tyrosinase gene associated with tyrosinase related oculocutaneous albinism (OCA1). Mutations in brief no. 204. Online.
Mutations of the human tyrosinase gene associated with tyrosinase related oculocutaneous albinism (OCA1). Mutations in brief no. 204. Online.
Human mutation (2000-02-12)
W S Oetting, J P Fryer, R A King
ABSTRACT
Mutations in the human tyrosinase gene produce tyrosinase-related oculocutaneous albinism (OCA1, MIM #203100). Tyrosinase is a copper containing enzyme and is responsible for catalyzing the rate limiting step in melanin biosynthesis, the hydroxylation of tyrosine to dopaquinone. We report 13 new mutations in the tyrosinase gene associated with OCA1A (without pigment) and OCA1B (with pigment) including 9 missense mutations (H19Q, R521, R77C, G97R, C289R, L312V, P313R, F340L and H404P), two nonsense mutations (W80X and R116X) and two frameshift mutations (53delG and 223 delG). Our previous work has defined clusters of missense mutations that appear to represent functional domains of the enzyme, and three of the missense mutations fall into these clusters including two (F340L and H404P) that flank the copper B bindng site and the missense mutation R52I that is located in the amino terminal end cluster of the protein. The G97R missense mutation is the first identified within the epidermal growth factor (EGF)-like sequence and the H19Q missense mutation alters the cleavage site of the signal peptide sequence. Mutational analysis can provide a definitive diagnosis of the type of OCA as well as help structure/function analysis.
Sigma-Aldrich
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid