Merck
  • 5'-Untranslated region of the tryptophan hydroxylase-2 gene harbors an asymmetric bidirectional promoter but not internal ribosome entry site in vitro.
5'-Untranslated region of the tryptophan hydroxylase-2 gene harbors an asymmetric bidirectional promoter but not internal ribosome entry site in vitro.
Gene (2009-04-07)
Guo-Lin Chen, Gregory M Miller
ABSTRACT
Tryptophan hydroxylase-2 (TPH2) catalyzes the synthesis of neuronal serotonin, a major neurotransmitter involved in many brain functions and psychiatric disorders. We have previously revealed a critical role of the human TPH2 (hTPH2) 5'-UTR in gene expression regulation. This study aimed to further characterize mechanism(s) by which the hTPH2 5'-UTR regulates gene expression. An internal ribosome entry site (IRES) activity in hTPH2 5'-UTR was suggested by the conventional bicistronic reporter assay; however, further stringent experiments, including in vitro translation, quantitative real-time PCR, Northern blot, ribonuclease protection assay, and monocistronic reporter assay, demonstrated that the hTPH2 5'-UTR harbors a bidirectional promoter, but not IRES, within its downstream segment (61-141). The antisense promoter is much stronger than the sense promoter, but the strength of both promoters are cell-line dependent, with the highest and lowest activities being observed in HEK-293T and SK-N-MC cells, respectively. In accordance with our previous findings, the upstream segment (1-60) of hTPH2 5'-UTR suppresses the neighboring promoter of both direction, independent of the cell line and its location in the 5'- or 3'-flanking regions of the gene. In summary, this study demonstrates that no IRES but an asymmetric bidirectional promoter is present in the downstream segment of hTPH2 5'-UTR, and this promoter is susceptible to a gene silencing effect caused by the upstream segment (1-60) of hTPH2 5'-UTR. Our findings point to the potential involvement of antisense transcription and non-coding RNA in the regulation of TPH2 gene expression.
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
SAFC
L-Methionine
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
D-Methionine, ≥98% (HPLC)
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material