- Polymorphisms in the human DNA polymerase beta gene.
Polymorphisms in the human DNA polymerase beta gene.
Human genetics (1995-04-01)
Y Dobashi, Y Kubota, T Shuin, S Torigoe, M Yao, M Hosaka
PMID7705833
ABSTRACT
Recently, evidence has accumulated that mutations in DNA repair genes might be associated with certain steps in carcinogenesis. The DNA polymerase beta gene is one of the DNA repair genes, and mutations in it have been detected in 83% of human colorectal cancers. To assess the involvement of polymerase beta gene mutations in the development of human prostate cancers, we performed sequence analyses of human DNA samples. Unexpectedly, we found six regions that were polymorphic. This information should be taken into consideration at the time of sequence analysis of the DNA polymerase beta gene.
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution