- THE PROPERTIES OF STREPTAVIDIN, A BIOTIN-BINDING PROTEIN PRODUCED BY STREPTOMYCETES.
THE PROPERTIES OF STREPTAVIDIN, A BIOTIN-BINDING PROTEIN PRODUCED BY STREPTOMYCETES.
Archives of biochemistry and biophysics (1964-07-20)
L CHAIET, F J WOLF
PMID14217155
Sigma-Aldrich
Streptavidin from Streptomyces avidinii, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Streptavidin from Streptomyces avidinii, essentially salt-free, lyophilized powder, ≥13 units/mg protein
Sigma-Aldrich
Streptavidin−FITC from Streptomyces avidinii, essentially salt-free, lyophilized powder, ≥5 units/mg protein
Sigma-Aldrich
Streptavidin from Streptomyces avidinii, affinity purified, lyophilized from 10 mM potassium phosphate, ≥13 U/mg protein
Sigma-Aldrich
Streptavidin−Alkaline Phosphatase from Streptomyces avidinii, lyophilized powder, ≥700 DEA units/mg protein