Merck
  • Expression of the CYP4F3 gene. tissue-specific splicing and alternative promoters generate high and low K(m) forms of leukotriene B(4) omega-hydroxylase.
Expression of the CYP4F3 gene. tissue-specific splicing and alternative promoters generate high and low K(m) forms of leukotriene B(4) omega-hydroxylase.
The Journal of biological chemistry (1999-07-20)
P Christmas, S R Ursino, J W Fox, R J Soberman
ABSTRACT
Cytochrome P450 4F3 (CYP4F3) catalyzes the inactivation of leukotriene B(4) by omega-oxidation in human neutrophils. To understand the regulation of CYP4F3 expression, we analyzed the CYP4F3 gene and cloned a novel isoform (CYP4F3B) that is expressed in fetal and adult liver, but not in neutrophils. The CYP4F3 gene contains 14 exons and 13 introns. The cDNAs for CYP4F3A (the neutrophil isoform) and CYP4F3B have identical coding regions, except that they contain exons 4 and 3, respectively. Both exons code for amino acids 66-114 but share only 27% identity. When expressed in COS-7 cells, the K(m) of CYP4F3B was determined to be 26-fold higher than the K(m) of CYP4F3A using leukotriene B(4) as a substrate. 5'-Rapid amplification of cDNA end studies reveal that the CYP4F3A and CYP4F3B transcripts have 5'-termini derived from different parts of the gene and are initiated from distinct transcription start sites located 519 and 71 base pairs (bp), respectively, from the ATG initiation codon. A consensus TATA box is located 27 bp upstream of the CYP4F3B transcription start site, and a TATA box-like sequence is located 23 bp upstream of the CYP4F3A transcription start site. The data indicate that the tissue-specific expression of functionally distinct CYP4F3 isoforms is regulated by alternative promoter usage and mutually exclusive exon splicing.
Sigma-Aldrich
Cytochrome P450 human, 4F3B isozyme microsomes, with P450 Reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Pricing and availability is not currently available.