Merck
  • Renal clearance of branched-chain L-amino and 2-oxo acids in maple syrup urine disease.
Renal clearance of branched-chain L-amino and 2-oxo acids in maple syrup urine disease.
Journal of inherited metabolic disease (1999-09-03)
P Schadewaldt, H W Hammen, A C Ott, U Wendel
ABSTRACT
In maple syrup urine disease (MSUD), branched-chain L-amino (BCAA) and 2-oxo acids (BCOA) accumulate in body fluids owing to an inherited deficiency of branched-chain 2-oxo acid dehydrogenase complex activity. In MSUD, little information is available on the significance of urinary disposal of branched-chain compounds. We examined the renal clearance of leucine, valine, isoleucine and alloisoleucine, and their corresponding 2-oxo acids 4-methyl-2-oxopentanoate (KIC), 3-methyl-2-oxobutanoate (KIV), (S)-(S-KMV), and (R)-3-methyl-2-oxopentanoate (R-KMV), using pairs of plasma and urine samples (n = 63) from 10 patients with classical MSUD. The fractional renal excretion of free BCAA was in the normal range (< 0.5%) and independent of the plasma concentrations. The excretion of bound (N-acylated) BCAA was normal and not significantly dependent on the BCAA plasma concentrations. The fractional renal excretion of BCOA was in the order KIC < KIV < R-KMV < or = S-KMV (range (%): KIC 0.1-25; KIV 0.14-21.3; S-KMV 0.26-24.6; R-KMV 0.1-35.9), significantly correlated with the KIC plasma concentrations, and generally higher than that of the related BCAA. The results show that the renal excretion of free BCAA as well as of the acylated derivatives is negligible. The renal excretion of BCOA, however, to some extent counteracts increases in BCAA concentrations and thus contributes to the lowering of total BCAA pools in MSUD.
Product Description
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
3-Methyl-2-oxopentanoic acid sodium salt, ≥98%
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
4-Methyl-2-oxovaleric acid, ≥98.0% (T)
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
SAFC
L-Valine
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
(S)-(−)-2-Hydroxyisocaproic acid, 98%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
(±)-3-Methyl-2-oxovaleric acid sodium salt
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Isoleucine
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
2-Hydroxy-3-methylbutyric acid, 99%
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material