Merck
  • Rate enhancement with high ratio of the monoalkylated product to the dialkylated product in the alkylation of the lithium enolate of 1-tetralone with reactive alkyl halides.
Rate enhancement with high ratio of the monoalkylated product to the dialkylated product in the alkylation of the lithium enolate of 1-tetralone with reactive alkyl halides.
Chemical & pharmaceutical bulletin (2000-10-25)
M Goto, K Akimoto, K Aoki, M Shindo, K Koga
ABSTRACT
The reactions of the lithium enolate of 1-tetralone with reactive alkyl halides were examined in the absence and in the presence of 3 eq of various ligands for the lithium. It is shown that the rates of the reactions are enhanced greatly in the presence a tetradentate amine (1,1,4,7,10,10-hexamethyltriethylenetetramine), and the ratio of the monoalkylated product to the dialkylated product is increased under shorter reaction times.
Sigma-Aldrich
β-Tetralone, 98%
Pricing and availability is not currently available.