Merck
  • Synergy between liquid chromatographic-pulsed amperometric detection and capillary-gas chromatographic methods for the detection of juice adulteration.
Synergy between liquid chromatographic-pulsed amperometric detection and capillary-gas chromatographic methods for the detection of juice adulteration.
Journal of AOAC International (2001-06-22)
D A Hammond
ABSTRACT
The use of liquid chromatography linked with pulsed amperometric detection, and capillary-gas chromatography to detect the adulteration of fruit juices is discussed. These are 2 complementary approaches which can be used to detect the addition of sugar syrups to juices at low levels.