Merck
Low-frequency collective chain dynamics in a model biomembrane.
Physical review. E, Statistical, nonlinear, and soft matter physics (2001-11-03)
K W Lee, C H Lee, D K Oh, J K Kang, C Eui Lee, J Kim
ABSTRACT
Proton NMR was employed as a probe for the collective hydrocarbon chain dynamics in decylammonium chloride (C10H21NH3Cl), a model biomembrane undergoing an irreversible structural phase transition sequence. Our rotating frame spin-lattice relaxation measurements revealed a low-frequency critical collective chain dynamics in the kHz regime, which is associated with the interdigitated to noninterdigitated chain configurational phase transition.
Sigma-Aldrich
Decylamine, 95%
Sigma-Aldrich
Decylamine, ≥99.0% (GC)