Merck
  • The N-terminal part of recombinant human tear lipocalin/von Ebner's gland protein confers cysteine proteinase inhibition depending on the presence of the entire cystatin-like sequence motifs.
The N-terminal part of recombinant human tear lipocalin/von Ebner's gland protein confers cysteine proteinase inhibition depending on the presence of the entire cystatin-like sequence motifs.
Biological chemistry (2001-12-01)
P Wojnar, W van't Hof, P Merschak, M Lechner, B Redl
ABSTRACT
Human Tear Lipocalin/von Ebner's gland protein (TL) is a member of the lipocalin superfamily. The protein is secreted by a number of serous glands and tissues and is overproduced under conditions of stress, infection and inflammation. In addition to its typical affinity for lipophilic ligands it was recently found to be able to inhibit cysteine proteinases [van't Hof et al., J. Biol. Chem. 272 (1997), 1837-1841], probably due to the presence of amino acid motifs resembling the papain binding domains of family 2 cystatins. In this work we have used a recombinant protein to confirm the results obtained with native TL. The inhibitory activity of the recombinant protein against papain was dependent on the ratio of papain and TL. At higher papain concentrations, the N-terminal sequence of TL was cleaved off by the protease, indicating that it can act in an inhibitor- or a substrate-like mode. This behaviour resembles that observed with certain chicken cystatin mutants. Using a recombinant TL mutant we found that the two Leu residues (Leu4-Leu5) contained within the first cystatin-like motif are absolutely essential for the inhibitory activity. These results were supported by experiments using a recombinant form of the corresponding pig von Ebner's gland protein (VEGp). This protein, which does not possess a fully conserved first cystatin-like motif, is unable to inhibit papain.
Sigma-Aldrich
Z-Phe-Arg 7-amido-4-methylcoumarin hydrochloride, kallikrein substrate