Merck
  • Propylene glycol monomethyl ether (PGME): inhalation toxicity and carcinogenicity in Fischer 344 rats and B6C3F1 mice.
Propylene glycol monomethyl ether (PGME): inhalation toxicity and carcinogenicity in Fischer 344 rats and B6C3F1 mice.
Toxicologic pathology (2002-10-10)
Pamela J Spencer, James W Crissman, William T Stott, Richard A Corley, Frank S Cieszlak, Alan M Schumann, Jerry F Hardisty
ABSTRACT
A series of inhalation studies with propylene glycol monomethyl ether (PGME) vapor were undertaken to characterize its subchronic toxicity in mice and chronic toxicity/oncogenicity in rats and mice. Groups of male and female Fischer 344 rats and B6C3F1 mice were exposed to 0, 300, 1,000, or 3,000 ppm vapor from 1 week to 2 years. Primary treatment-related effects included: initial sedation of animals exposed to 3,000 ppm and its subsequent resolution correlating with induction of hepatic mixed function oxidase activity and S-phase DNA synthesis; elevated mortality in high-exposure male rats and mice (chronic study); elevated deposition of alpha2u-globulin (alpha2U-G) and associated nephropathy and S-phase DNA synthesis in male rat kidneys; accelerated atrophy of the adrenal gland X-zone in female mice (subchronic study only); and increased occurrence and/or severity of eosinophilic foci of altered hepatocytes in male rats. No toxicologically relevant statistically significant increases in neoplasia occurred in either species. A numerical increase in the incidence of kidney adenomas occurred in intermediate-exposure male rats; however, the association with alpha2U-G nephropathy, a male rat specific effect, indicated a lack of relevance for human risk assessment.
Sigma-Aldrich
1-Methoxy-2-propanol, ≥99.5%
Supelco
1-Methoxy-2-propanol, analytical standard