Merck
  • A direct continuous pH-spectrophotometric assay for arginine kinase activity.
A direct continuous pH-spectrophotometric assay for arginine kinase activity.
Protein and peptide letters (2003-01-30)
Z Yu, J Pan, H M Zhou
ABSTRACT
A direct and continuous spectrophotometric method was developed for determining arginine kinase (phosphoarginine synthesis) activity. Protons are produced during the phosphoarginine synthesis course, so adding the complex acid-base indicator to this solution and monitoring the decrease of absorbance of the solution at 575 nm will follow the arginine kinase activity. For this condition, one activity unit of arginine kinase was defined as 1 micromol H+ produced in 1 minute in the enzymatic reaction catalyzed by arginine kinase.
Sigma-Aldrich
Cresol red, indicator grade, Dye content 95 %
Sigma-Aldrich
Cresol Red sodium salt, indicator grade
Supelco
Thymolphthalein, ACS reagent
Sigma-Aldrich
Thymolphthalein, ACS reagent, Dye content 95 %
Sigma-Aldrich
Thymol Blue, ACS reagent
Sigma-Aldrich
Thymol Blue sodium salt, ACS reagent, Dye content 95 %