Merck
  • A new method for enzymatic preparation of isopentenyladenine-type and trans-zeatin-type cytokinins with radioisotope-labeling.
A new method for enzymatic preparation of isopentenyladenine-type and trans-zeatin-type cytokinins with radioisotope-labeling.
Journal of plant research (2003-05-03)
Kentaro Takei, Yasumasa Dekishima, Tadashi Eguchi, Tomoyuki Yamaya, Hitoshi Sakakibara
ABSTRACT
We describe a new enzymatic reaction method for the preparation of the radioisotope-labeled cytokinins isopentenyladenine (iP), trans-zeatin (tZ), and their ribosides. The method is based on the three enzyme activities of an adenylate isopentenyltransferase (IPT; EC 2.5.1.27) from Arabidopsis thaliana, an alkaline phosphatase (EC 3.1.3.1) from calf intestine, and a purine-nucleoside phosphorylase (EC 2.4.2.1) from Escherichia coli. The A. thaliana IPT, AtIPT7, utilized both dimethylallyldiphosphate and 4-hydroxy-3-methyl-2-( E)-butenyl diphosphate as isoprenoid donors. The dual specificity of the substrates enabled us to produce iP-type and tZ-type cytokinins separately in the same system simply by switching the substrates. Our method affords a much higher yield of the labeled products than the chemical reaction methods previously used. These labeled compounds will be useful tools for cytokinin research, such as receptor-ligand assays and cell metabolism studies.
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, BioReagent, suitable for plant cell culture, 1 mg/mL
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, suitable for plant cell culture, BioReagent, ≥90%
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, suitable for plant cell culture, BioReagent, ≥98.5%