Merck
Fragrance material review on linalyl formate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2003-06-14)
C S Letizia, J Cocchiara, J Lalko, A M Api
ABSTRACT
A toxicologic and dermatologic review of linalyl formate when used a a fragrance ingredient is presented.