Merck
Synthesis of porritoxin.
The Journal of organic chemistry (2004-04-03)
Ivan Cornella, T Ross Kelly
ABSTRACT
A total synthesis following the sequence in Scheme 1 confirms that porritoxin possesses revised structure 3, not the originally assigned 1. A key reaction was the use of iron pentacarbonyl to formylate an aryllithium when DMF and methyl formate proved insufficiently reactive.
Sigma-Aldrich
Methyl formate, anhydrous, 99%
Supelco
Methyl formate, analytical standard
Sigma-Aldrich
Methyl formate, reagent grade, 97%
Sigma-Aldrich
Methyl formate, spectrophotometric grade, ≥98%