Merck
  • Major factors modulating the serum oxalic acid level in hemodialysis patients.
Major factors modulating the serum oxalic acid level in hemodialysis patients.
Frontiers in bioscience : a journal and virtual library (2004-09-09)
Yoshihide Ogawa, Noriko Machida, Masahide Jahana, Munehisa Gakiya, Yoshiaki Chinen, Masami Oda, Makoto Morozumi, Kimio Sugaya
ABSTRACT
Ascorbic acid overload and vitamin B6 deficiency have been implicated in the development of hyperoxalemia in dialysis patients, but there is still disagreement about this. Hemodialysis patients who are exposed long-term hyperoxalemia may develop secondary oxalosis with an increased risk of cardiac, vascular, and bone disease, and thus may benefit from maintaining a low serum oxalic acid level. In 452 hemodialysis patients, the serum level of oxalic acid was 47.2 +/- 22.9 micromol /l before and 16.9 +/- 10.5 micromol/l after a 4-hour dialysis session, while the ascorbic acid levels were 39.0 +/- 92.7 micromol/l and 6.5 +/- 18.6 micromol/l, the glycolic acid levels were 7.3 +/- 10.1 micromol/l and 0.6 +/- 2.3 micromol/l, and the citric acid levels were 141.3 +/- 54.7 micromol/l and 117.6 +/- 37.2 micromol/l, respectively. Most patients (65.3 percent) had low serum ascorbic acid levels (less than 10 micromol/l) before hemodialysis. The serum level of oxalic acid [Ox] showed a significant positive correlation with the levels of ascorbic acid [AA], glycolic acid [Gly], and creatinine [Cre]: [Ox] = 21.711 + 0.181 x [AA] + 0.174 x [Gly] + 0.171 x [Cre], (all micromol/l, p less than 0.05). In 124 dialysis patients, the 4-pyridoxic acid level was 8.9 +/- 19.6 micromol /l before and 3.9 +/- 8.8 micromol/l after dialysis, and it was not correlated with oxalic acid or glycolic acid. Most dialysis patients (65.3 percent) had low serum levels of ascorbic acid, but a subgroup of patients (12 percent) had high serum ascorbic acid levels (more than 100 micromol/l) associated with hyperoxalemia (88.2 +/- 24.5 micromol/l). High-dose vitamin C supplementation may aggravate hyperoxalemia in hemodialysis patients, so attention should be paid to avoiding this risk.
Product Description
Supelco
Glycolic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
SAFC
L-Proline
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
L-Proline, from non-animal source, meets EP, USP testing specifications, suitable for cell culture