Merck
  • Determination of microcystin variants and related peptides present in a water bloom of Planktothrix (Oscillatoria) rubescens in a Spanish drinking water reservoir by LC/ESI-MS.
Determination of microcystin variants and related peptides present in a water bloom of Planktothrix (Oscillatoria) rubescens in a Spanish drinking water reservoir by LC/ESI-MS.
Toxicon : official journal of the International Society on Toxinology (2004-11-09)
Mónica Barco, Cintia Flores, Josep Rivera, Josep Caixach
ABSTRACT
A water bloom of Planktothrix (Oscillatoria) rubescens was observed in a drinking water reservoir in central Spain in 2003. Microcystins where analysed by LC/ESI-MS in 21 samples collected from this reservoir in five different days between March and May. A demethylated variant of microcystin-RR was identified as the major microcystin in most samples. Trace levels of microcystin-LR, -RR and -YR were detected in some samples. Four less common microcystins, with [M+H]+ ions at m/z 960, 981, 1045 and 1053, were also found. Total extracellular microcystin concentration varied from 0.010 to 19.126 microg l(-1). Furthermore, anabaenopeptins B and F as well as Oscillamide Y were also identified in these samples.
Product Description
Supelco
Microcystin-YR solution, 10 μg/mL in methanol, analytical standard