Merck
  • Conformational studies of resin-bound vancomycin and the complex of vancomycin and Ac2-L-Lys-D-Ala-D-Ala.
Conformational studies of resin-bound vancomycin and the complex of vancomycin and Ac2-L-Lys-D-Ala-D-Ala.
Journal of combinatorial chemistry (2005-01-11)
Nian-Huan Yao, Wen-Yi He, Kit S Lam, Gang Liu
ABSTRACT
The molecular target of vancomycin, a commonly used glycopeptide antibiotic, is the D-Ala-D-Ala dipeptide subunit on the bacterial cell wall. The molecular basis of interaction between vancomycin and D-Ala-D-Ala in solution is well-known. However, there is no structural data on vancomycin, and its interaction with D-Ala-D-Ala when the drug is tethered to a solid support. In this Article, vancomycin was directly coupled onto TentaGel or PEGA resin through its C terminus. High-resolution magic angle spinning NMR studies indicated that conformation of PEGA bead-bound vancomycin is identical to that of the free drug. Broadening and shifts of the same proton resonances were observed in solution-phase vancomycin or PEGA-bound vancomycin when complexed with Ac(2)-L-Lys-D-Ala-D-Ala. This study demonstrates that bead-bound molecules can behave the same as solution-phase molecules in terms of molecular interaction with its target molecule, thus validating the on-bead screening approach of the "one-bead-one-compound" combinatorial library method.
Sigma-Aldrich
Nα,Nε-Diacetyl-Lys-D-Ala-D-Ala, carboxypeptidase substrate
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), average Mn 3400
Sigma-Aldrich
Poly(ethylene glycol) diamine, average Mn 6,000
Sigma-Aldrich
Poly(ethylene glycol) diamine, average Mn 3,000
Sigma-Aldrich
Poly(ethylene glycol) diamine, average Mn 10,000
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 2,000
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 20,000
Sigma-Aldrich
Poly(ethylene glycol) diamine, average Mn 2,000
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 6,000
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 3,000
Sigma-Aldrich
Poly(ethylene glycol) bis(amine), Mw 10,000