Merck
  • Process modeling of the lipase-catalyzed dynamic kinetic resolution of (R, S)-suprofen 2,2,2-trifluoroethyl thioester in a hollow-fiber membrane.
Process modeling of the lipase-catalyzed dynamic kinetic resolution of (R, S)-suprofen 2,2,2-trifluoroethyl thioester in a hollow-fiber membrane.
Bioprocess and biosystems engineering (2005-01-13)
L W Wang, Y C Cheng, S W Tsai
ABSTRACT
A Candida rugosa lipase immobilized on polypropylene powder was employed as the biocatalyst for the enantioselective hydrolysis of (R, S)-suprofen 2,2,2-trifluorothioester in cyclohexane, in which trioctylamine was added as the catalyst to perform in situ racemization of the remaining (R)-thioester. A hollow-fiber membrane was also integrated with the dynamic kinetic resolution process in order to continuously extract the desired (S)-suprofen into an aqueous solution containing NaOH. A kinetic model for the whole process (operating in batch and feed-batch modes) was developed, in which enzymatic hydrolysis and deactivation, lipase activation, racemization and non-enantioselective hydrolysis of the substrate by trioctylamine, and reactive extraction of (R)- and (S)-suprofen into the aqueous phase in the membrane were considered. Theoretical predictions from the model for the time-course variations of substrate and product concentrations in each phase were compared with experimental data.
Sigma-Aldrich
Trioctylamine, 98%