Merck
  • Herpesvirus entry mediator ligand (HVEM-L), a novel ligand for HVEM/TR2, stimulates proliferation of T cells and inhibits HT29 cell growth.
Herpesvirus entry mediator ligand (HVEM-L), a novel ligand for HVEM/TR2, stimulates proliferation of T cells and inhibits HT29 cell growth.
The Journal of biological chemistry (1998-10-09)
J A Harrop, P C McDonnell, M Brigham-Burke, S D Lyn, J Minton, K B Tan, K Dede, J Spampanato, C Silverman, P Hensley, R DiPrinzio, J G Emery, K Deen, C Eichman, M Chabot-Fletcher, A Truneh, P R Young
ABSTRACT
Herpesvirus entry mediator (HVEM), a member of the tumor necrosis factor (TNF) receptor family, mediates herpesvirus entry into cells during infection. Upon overexpression, HVEM activates NF-kappaB and AP-1 through a TNF receptor-associated factor (TRAF)-mediated mechanism. Using an HVEM-Fc fusion protein, we screened soluble forms of novel TNF-related proteins derived from an expressed sequence tag data base. One of these, which we designated HVEM-L, specifically bound to HVEM-Fc with an affinity of 44 nM. This association was confirmed with soluble and membrane forms of both receptor and ligand. HVEM-L mRNA is expressed in spleen, lymph nodes, macrophages, and T cells and encodes a 240-amino acid protein. A soluble, secreted form of the protein stimulates proliferation of T lymphocytes during allogeneic responses, inhibits HT-29 cell growth, and weakly stimulates NF-kappaB-dependent transcription.
Sigma-Aldrich
LIGHT from mouse, recombinant, expressed in E. coli, ≥96% (SDS-PAGE), ≥96% (HPLC), suitable for cell culture