Merck
  • Chemiluminescence in molecular recognition: base-induced decomposition of optically active dioxetanes bearing a bisnaphthol moiety with a complex of optically active crown ether-potassium tert-butoxide.
Chemiluminescence in molecular recognition: base-induced decomposition of optically active dioxetanes bearing a bisnaphthol moiety with a complex of optically active crown ether-potassium tert-butoxide.
Chemical communications (Cambridge, England) (2005-02-03)
Masakatsu Matsumoto, Koji Hamaoka, Yuji Takashima, Mizuki Yokokawa, Kazutaka Yamada, Nobuko Watanabe, Hisako K Ijuin
ABSTRACT
Four optically pure isomers of dioxetane (1) were decomposed by the action of the complex of an optically active crown ether with potassium tert-butoxide to afford light with lambda(max)CL and gave four different spectrum shapes.
Sigma-Aldrich
Potassium tert-butoxide, reagent grade, ≥98%
Sigma-Aldrich
Potassium tert-butoxide, sublimed grade, 99.99% trace metals basis
Sigma-Aldrich
Potassium tert-butoxide solution, 1.0 M in THF