Merck
  • 3-nitropropionic acid-induced hydrogen peroxide, mitochondrial DNA damage, and cell death are attenuated by Bcl-2 overexpression in PC12 cells.
3-nitropropionic acid-induced hydrogen peroxide, mitochondrial DNA damage, and cell death are attenuated by Bcl-2 overexpression in PC12 cells.
Brain research. Molecular brain research (2005-02-16)
Bhaskar S Mandavilli, Istvan Boldogh, Bennett Van Houten
ABSTRACT
3-nitropropionic acid (3-NPA), a complex II inhibitor of the electron transport chain, causes Huntington disease-like symptoms after administration into animals. However, primary mechanisms of cell death are not clearly understood. This study tested the hypothesis that 3-NPA leads to the generation of reactive oxygen species (ROS), mitochondrial DNA damage, and loss of mitochondrial function. Amplex red and horseradish peroxidase were used to accurately measure the amount of H2O2, and showed that PC12 cells treated with 3-NPA (4 mM) lead to the production of hydrogen peroxide (1 nmol/10(6) cells/h). This amount of 3-NPA also leads to a rapid decline of ATP levels. There was time- and dose-dependent mitochondrial DNA damage following 3-NPA treatment. Overexpression of the proto-oncogene bcl-2 protects cells from apoptosis induced by various stimuli. Overexpression of Bcl-2 leads to almost threefold higher levels of ATP and also decreased the 3-NPA-mediated induction of hydrogen peroxide by over 50%. Bcl-2-overexpressing PC12 cells were also protected from mitochondrial DNA damage. These data show that ROS production followed by mitochondrial DNA damage is the primary event in 3-NPA toxicity, and Bcl-2 protects PC12 cells from 3-NPA toxicity by preventing mitochondrial DNA damage.
Product Description
Supelco
Dihydrotachysterol, analytical standard