Merck
  • Main compounds responsible for off-odour of strawberries infected by Phytophthora cactorum.
Main compounds responsible for off-odour of strawberries infected by Phytophthora cactorum.
Letters in applied microbiology (2005-03-09)
H H Jeleń, J Krawczyk, T O Larsen, A Jarosz, B Gołebniak
ABSTRACT
Volatile compounds present in strawberries infected with Phytophthora cactorum, especially those responsible for the characteristic off-odour of such fruits were the subject of this study. Six strawberry varieties (Redgauntlet, Selva, Korona, Tenira, Real, Pegasus) inoculated with P. cactorum strain (PC-5), isolated from naturally infected fruit and one variety inoculated with 15 strains of P. cactorum in the laboratory were analysed. All the samples had a distinct, to a various degree, off-odour reminiscent of watercolour paint with phenolic notes. Volatile compounds were isolated by solid phase microextraction and simultaneous distillation extraction methods. To detect compounds responsible for the characteristic off-odour, gas chromatography-olfactometry was used. Two compounds were found to be responsible for the characteristic off-odour of strawberries infected by P. cactorum: 4-ethyl phenol and 4-ethyl-2-metoxy phenol (4-ethyl guaiacol). The content of these compounds in infected varieties ranged from 1.12 to 22.56 mg kg(-1) and 0.14-1.05 mg kg(-1) respectively. Other volatile compounds, not detected in noninoculated sound strawberries, were also identified: camphene, 1-octene-3-ol, 3-octanone, o-cymene, phenyl methanol, cis-linaloloxide, nonanal, phenyl ethyl alcohol, 2-undecanone and alpha-muurolene. Volatile compounds responsible for the characteristic off-odour of strawberries infected with P. cactorum were identified. Also compounds produced as a result of P. cactorum growth on strawberry fruit were characterized.
Sigma-Aldrich
4-Ethylguaiacol, natural, FG
Supelco
3-Octanone, analytical standard
Sigma-Aldrich
3-Octanone, ≥98%, FG
Sigma-Aldrich
4-Ethylguaiacol, ≥98%, FCC, FG
Sigma-Aldrich
3-Octanone, ≥98%
Sigma-Aldrich
Nonanal, 95%
Supelco
Nonanal, analytical standard
Sigma-Aldrich
Nonanal, ≥95%, FCC