Merck
  • Fluorinated phenylcyclopropylamines. Part 4: effects of aryl substituents and stereochemistry on the inhibition of monoamine oxidases by 1-aryl-2-fluoro-cyclopropylamines.
Fluorinated phenylcyclopropylamines. Part 4: effects of aryl substituents and stereochemistry on the inhibition of monoamine oxidases by 1-aryl-2-fluoro-cyclopropylamines.
Bioorganic & medicinal chemistry (2005-03-10)
Song Ye, Shinichi Yoshida, Roland Fröhlich, Günter Haufe, Kenneth L Kirk
ABSTRACT
A series of para-ring-substituted (E)- and (Z)-1-aryl-2-fluorocyclopropylamines were examined as inhibitors of recombinant human liver monoamine oxidase A (MAO A) and B (MAO B). Unlike the parent 1-phenylcyclopropylamine, which is a selective inhibitor of MAO B, both (E)- and (Z)-diastereomers of derivatives having fluorine at the 2-position of the cyclopropane ring were potent and selective irreversible inhibitors of MAO A. Both electron releasing groups (Me, OMe) and electron attracting groups (Cl, F) substituted in the para-position caused a modest increase in activity. Geminal difluoro-substitution caused a loss of potency of 100-fold compared to either (E)- or (Z)-monofluorinated analogue. Surprisingly, (1S,2R)-2-fluoro-1-phenylcyclopropylamine and the (1R,2S)-enantiomer were essential equally potent as inhibitors of MAO A and MAO B. None of the tested 1-aryl-2-fluorocyclopropylamines exhibited significant inhibition of tyramine oxidase.
Sigma-Aldrich
Cyclopropylamine, 98%