Merck
Solubilization of unconjugated bilirubin by bile salts.
Experientia (1979-06-15)
U Wosiewitz, S Schroebler
PMID38133
ABSTRACT
Freshly precipitated unconjugated bilirubin (UCB) is solubilized rapidly and to a large extent by the sodium salts of di- and trihydroxy bile acids. The solubilization effect depending on bile salt concentration, pH and ionic strength is based on micellar mechanisms.
Sigma-Aldrich
Sodium taurocholate hydrate, ≥97.0% (TLC)